Z-DEVD-pNA
Catalog # ENZOALX260048M001
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:1 mg
- Seq:02
- Protein/peptide name:Z-DEVD-pNA
- Purity:>99% (HPLC)
- Formulation:Lyophilized
Specifications
About this item
Caspase substrate
Chromogenic substrate for caspase-3 with increased cell permeability. Also substrate for caspase-1, -4 and -7. λmax of pNA is 400 nm. Similar to Ac-DEVD-pNA except the N-terminus is blocked by a benzyloxycarbonyl (Z) moiety which may increase cell permeability.