Human recombinant Elafin
Catalog # ENZOALX201240C100
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Species:Human
- Seq:02
- Biological activity:Inhibits human leukocyte elastase (Ki=0,17 nM) and human proteinase 3 (Ki=0,42 nM).
- Reconstitution instructions:Reconstitute in sterile distilled water. Further dilutions should be made in aqueous buffers, pH 4,0 - 8,0.
- Protein/peptide name:Elafin
- Purity:>90% (HPLC, SDS-PAGE, MS)
- Formulation:Lyophilised
Specifications
About this item
Produced in yeast.
Belongs to the chelonianin family of protease inhibitors and is involved in the physiological control of neutrophil serine proteinases, where it targets NE and proteinase 3. Shows antiprotease, antibacterial and antiinflammatory activity and primes innate immunity.