Recombinant GrpE (from E. coli)
Catalog # ENZOALX201145C025
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:25 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Seq:02
- Protein/peptide name:GrpE
- Purity:>99%
- Concentration:2,15 mg/ml
- Formulation:Liquid. In 30 mM HEPES/KOH, pH 7,0, containing 10 mM KCl and 5% glycerol.
Specifications
About this item
Produced in E. coli.