Recombinant DnaJ (from E. coli)
Catalog # ENZOALX201144C025
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:25 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Seq:02
- Protein/peptide name:DnaJ
- Purity:>99%
- Concentration:4,3 mg/ml
- Formulation:Liquid. In 30 mM HEPES/KOH, pH 8,2, containing 120 mM KCl, 15 mM NaCl and 4% glycerol.
Specifications
About this item
Produced in E. coli.