Human Recombinant Granzyme A (from E. coli)
Catalog # ENZOALX201118C010
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Protein/peptide name:Granzyme A
- Purity:>98% (SDS-PAGE)
- Concentration:0,2 mg/ml
- Formulation:Liquid. In 0,05 M MES, pH 6,0, containing 10% glycerol and ~0,52 M sodium chloride. Contains no preservatives.
Specifications
About this item
Produced in E. coli. Active human granzyme A.