Human recombinant MMP-14 (catalytic domain) (89-265) (from E. coli)
Catalog # ENZOALX201098C010
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Protein/peptide name:MMP-14 (catalytic domain) (89-265)
- Purity:>95% (SDS-PAGE)
- Concentration:0,2 mg/ml
- Formulation:Liquid. In 50 mM TRIS-HCl, pH 7,5, 150 mM NaCl, 5 mM CaCl₂.
Specifications
About this item
Produced in E. coli. Mature human MMP-14 (aa 89-265).