Order Entry
Czech Republic
ContactUsLinkComponent
MALP-2 (BULK)
MALP-2 (BULK)
Catalog # ENZOALX162034C500
Supplier:  Enzo Life Sciences
CAS Number:  
MALP-2 (BULK)
Catalog # ENZOALX162034C500
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    500 µG
  • Seq:
    02
  • Biological activity:
    Stimulates macrophages, monocytes and other TLR2 and TLR6 expressing cells in vitro at pg/ml concentrations (e.g. mouse macrophages: 50pg/ml; fibroblasts: 2ng/ml) to release various mediators. Human monocytes require about 10-100x higher concentrations fo
  • Protein/peptide name:
    MALP-2 (BULK)
  • Concentration:
    1 mg/ml
  • Formulation:
    Liquid. Pyrogen-free, sterile solution in 33% (v/v) 2-propanol/water.
  • Purification:
    HPLC purified

Specifications

About this item

TLR activator

MALP-2 was originally isolated from Mycoplasma fermentans. This MALP-2 corresponds to the originally isolated isomer, which expresses potent endotoxin-like activity and approaches in certain experimental systems the toxicity of LPS. For description of the stereochemistry of MALP-2. MALP-2 signalling, unlike that of LPS, is not transduced via TLR4, but is induced via TLR2 and TLR6 signalling. For more information about MALP-2 see www.malp-research.de. BULK size (500 µg) of biologically active, pyrogen-free and sterile MALP-2 for in vitro and animal in vivo studies.