Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Angiotensin III
Angiotensin III
Angiotensin III
Catalog # ENZOALX151043M005
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Expand image
Angiotensin III
Catalog # ENZOALX151043M005
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Specifications
Pk:
5 mg
Seq:
02
Protein/peptide name:
Angiotensin III
Specifications
About this item
Product Details
Product Details