Order Entry
Czech Republic
ContactUsLinkComponent
Human recombinant HSF1 (phosphorylated) (from E. coli), HIS tag
Human recombinant HSF1 (phosphorylated) (from E. coli), HIS tag
Catalog # ENZOADISPP902D
Supplier:  Enzo Life Sciences
CAS Number:  
Human recombinant HSF1 (phosphorylated) (from E. coli), HIS tag
Catalog # ENZOADISPP902D
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    02
  • Tag sequence:
    HIS Tag
  • Protein/peptide name:
    HSF1 (phosphorylated)
  • Purity:
    >90% (~60% full-length with trace upstream kinase)
  • Formulation:
    Liquid. In 1x TRIS buffered saline containing 5% glycerol.
  • Purification:
    Purified by multi-step chromatography.

Specifications

About this item

Produced in E. coli. Human phosphorylated HSF1 is fused at the N-terminus to a His-tag.