Human Recombinant Grp75/Mortalin (from E. coli)
Produced in E. coli.
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Protein synonyms:Mortalin
- Protein/peptide name:Grp75
- Purity:-
- Formulation:-
- Purification:-
Frequently Bought Together