Human recombinant HSP70B' (from E. coli)
Catalog # ENZOADISPP762E
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Protein/peptide name:HSP70B
- Formulation:Liquid, in Dulbecco's PBS, pH 7,2, containing 8,1 mM sodium phosphate, 7H₂O, 1,5 mM potassium phoshate, 2,7 mM potassium chloride, and 137 mM sodium chloride
- Purification:Purified by multi-step chromatography
Specifications
About this item
Produced in E. coli.