Human recombinant HSP60 (from E. coli)
Catalog # ENZOADINSP540E
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Protein/peptide name:HSP60
- Purity:>90% (SDS-PAGE; Western blot)
- Formulation:Liquid, In Dulbecco's PBS
- Purification:Purified by multi-step chromatography
Specifications
About this item
Produced in E.coli.