Human recombinant HSP27 (from E. coli)
Dodavatel: Enzo Life Sciences
Certifikáty
O tomto produktu
Produced in E.coli.
Specifikace
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Protein/peptide name:HSP27
- Endotoxin level:Low
- Purity:>90% (SDS-PAGE; Western blot)
- Formulation:Liquid, In Dulbecco's PBS
- Purification:Purified by multi-step chromatography
Specifikace
Frequently Bought Together