Order Entry
Czech Republic
ContactUsLinkComponent
Nuclear-ID™ Blue/Red cell viability reagent (GFP-Certified™)
Nuclear-ID™ Blue/Red cell viability reagent (GFP-Certified™)
Catalog # ENZOENZ53005C100
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  
Nuclear-ID™ Blue/Red cell viability reagent (GFP-Certified™)
Catalog # ENZOENZ53005C100
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  

Specifications

  • Pk:
    100 µl
  • Host:
  • Description:
    Nuclear-ID™ Blue/red cell viability reagent (GFP-Certified™)
  • Seq:
    02
  • Environmentally Preferable:
  • Purity:
    ≥93% (HPLC)

Specifications

About this item

Cell viability dye

The Nuclear-ID™ Blue/Red cell viability reagent (GFP-Certified™) (Prod. No. ENZ-53005) is a mixture of a blue fluorescent cell-permeable nucleic acid dye and a red fluorescent cell-impermeable nucleic acid dye that is suited for staining dead nuclei. The staining pattern arising from the simultaneous combination of these two dyes permits determination of live and dead cell populations by fluorescence/confocal microscopy. The reagent, supplied as a 1000x solution, is sufficient for 1000 microscopy assays. The single-tube format makes this cell viability reagent easy to use. It leaves the cytoplasm unstained, potentially allowing visualization of other cell markers. This reagent is specificially designed for use with GFP-expressing cell lines.

Ordering information: Quantity: 100 μl (for 1000 microscopy assays).