Order Entry
Czech Republic
ContactUsLinkComponent
AMPIGENE® dNTP Mix
AMPIGENE® dNTP Mix
Catalog # ENZOENZNUC1020500
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
AMPIGENE® dNTP Mix
Catalog # ENZOENZNUC1020500
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    500 µl
  • Description:
    AMPIGENE® dNTP mix
  • Seq:
    02
  • Purity:
    ≥99%
  • Concentration:
    10 mM

Specifications

About this item

Ultra-pure, stable, and suitable for a variety of PCR applications.

AMPIGENE® dNTP Mix is an ultrapure mix of deoxyribonucleotide triphosphates to use as building blocks for a wide variety of PCR applications. It can withstand multiple freeze-thaw cycles and shown to retain 95% dNTPs in triphosphate form after 5 weeks at room temperature. AMPIGENE® dNTP Mix has been tested on many molecular biology applications including standard PCR, fast PCR, 1-step PCR, high-fidelity PCR, real-time PCR, lon-range PCR, MDA, RDA, LAMP, and DNA sequencing.