Order Entry
Czech Republic
ContactUsLinkComponent
PGE2 FPIA kit
PGE2 FPIA kit
Catalog # ENZOADI920001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
PGE2 FPIA kit
Catalog # ENZOADI920001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    100 Tests
  • Target protein:
    PGE2
  • Description:
    PGE2 FPIA kit
  • Seq:
    02
  • Sensitivity:
    684 pg/ml (range 1,562 - 100,000 pg/ml)

Specifications

About this item

For the quantitative determination of PGE2 in culture supernatants from any species.

The PGE2 FPIA kit is a homogeneous mix-and-read fluorescence polarization enzyme immunoassay kit with. Results are obtained within just 30 minutes and the generated signal is stable for up to 20 hours. The PGE2 FPIA kit is ideal for high throughput screening (HTS) applications. A convenient homogeneous mix-and-read assay that is amendable to miniaturization and automation, this kit has a robust Z factor >0.8.

Ordering information: Contains Conjugate concentrate, Antibody, Assay buffer concentrate, Standard.