ContactUsLinkComponent
AEC Peroxidase substrate kit
AEC Peroxidase substrate kit
Catalog # ENZOENZ43825
Supplier:  Enzo Life Sciences
CAS Number:  
AEC Peroxidase substrate kit
Catalog # ENZOENZ43825
Supplier:  Enzo Life Sciences
CAS Number:  
Specifications
  • Pk:
    300 mL
  • Description:
    AEC peroxidase substrate kit
  • Seq:
    02
Specifications
About this item
The AEC Peroxidase Substrate Kit contains reagents for visualization of peroxidase reactions using Aminoethylcarbazole (AEC). In the presence of H2O2, peroxidases catalyze the oxidation of AEC into a brick red precipitate. The supplied reagents are sufficient for preparation of 300 ml of ready-to-use color reaction mixture.
Ordering information: Contents: Aminoethylcarbazole, 6 ml 20 mg/ml in dimethylformamide 8X Reaction Buffer/Substrate Reagent, 40 ml Hydrogen peroxide in acetate buffer with stabilizers and enhancers Mixing Bottle, 1 15 ml bottle for preparation of color reaction mixture