Matrix Metalloproteinase-1 (MMP-1) colorimetric drug discovery kit
Catalog # ENZOBMLAK4040001
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:1 KIT
- Description:Matrix Metalloproteinase-1 (MMP-1) colorimetric drug discovery kit
- Seq:02
- Storage conditions:−80 °C
- Application:Colorimetric detection, HTS activity assay
- Size:1 kit
Specifications
About this item
A QuantiZyme™ Assay System. The MMP-1 Colorimetric Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-1 (MMP-1, interstitial collagenase, fibroblast collagenase) using a thiopeptide as a chromogenic substrate.
Delivery: Kit includes MMP-1 Enzyme (human) (recombinant), Substrate, inhibitor, Assay Buffer and 96-well Microplate.