BNP fragment ELISA kit, grow cells
Katalogové číslo ENZOENZKIT1520001
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:1 KIT
- Assay Type:Competitive
- Description:BNP fragment ELISA kit, grow cells
- Seq:01
- Sensitivity:171 pmol/L (0-6400 pmol/L)
Specifications
About this item
Sensitive BNP fragment ELISA kit enabling detection of BNP fragment in serum and plasma.
- Quantitative determination of human BNP fragment
- Colorimetric, competitive enzyme immunoassay
For the quantitative determination of human BNP fragment in serum and plasma. The BNP Fragment ELISA kit is a colorimetric competitive enzyme immunoassay kit.