Order Entry
Czech Republic
ContactUsLinkComponent
 
Insulin ELISA kit
Catalog # ENZOENZKIT1410001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Insulin ELISA kit
Catalog # ENZOENZKIT1410001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Pig
    Human
  • Target protein:
    Insulin
  • Description:
    Insulin ELISA kit, grow cells
  • Seq:
    01
  • Sample type:
    Serum, plasma, tissue culture media
  • Cross reactivity:
    No cross-reactivity to similar family molecules (proinsulin)
  • Detection range:
    15,6 - 500 pg/ml
  • Sensitivity:
    15,5 pg/ml
  • Regulatory status:
    RUO
  • Tests per kit:
    96 wells

Specifications

About this item

Highly sensitive Insulin ELISA kit enabling detection of insulin in serum, plasma, tissue culture media in just 3 hours.

  • Highly sensitive measurement of insulin, detecting as little as 21,6 pg/ml (0,622 µIU/ml)
  • Negligible cross reactivity with proinsulin
  • High throughput format with results in 3 hours for up to 40 samples in duplicate
  • Fully quantitative results that surpass semi-quantitative Western blot analysis

For the quantitative determination of insulin in serum, plasma, and tissue culture media. The Insulin ELISA kit is a colorimetric immunometric immunoassay kit with results in 3 hours.

Ordering information: Contains Microtiter plate, Assay Buffer 13, Standard, Wash Buffer Concentrate, Antibody, Antibody Diluent, Conjugate, Substrate, and Stop Solution.