Order Entry
Czech Republic
ContactUsLinkComponent
TGF-α ELISA kit, Enzo Life Sciences
TGF-α ELISA kit, Enzo Life Sciences
Catalog # ENZOENZKIT1320001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
TGF-α ELISA kit, Enzo Life Sciences
Catalog # ENZOENZKIT1320001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    TGF-α ELISA kit, grow cells
  • Seq:
    01
  • Sensitivity:
    9,375 pg/ml

Specifications

About this item

Highly sensitive ELISA kit enabling detection of TGF-α in 3,5 hours.

  • Sensitive measurement of TGF-α, detecting as little as 9,375 pg/ml
  • Negligible reactivity from similar growth factors
  • High throughput format with results in 3,5 hours
  • Fully quantitative results that surpass semi-quantitative Western blot analysis

For the quantitative determination of TGF-α in serum, plasma, cell culture supernatant, tissue homogenates in human samples.

The TGF-α ELISA kit is a colorimetric immunometric immunoassay kit with results in 3,5 hours.

Ordering information: Contains Microtiter Plate, Standard, Detection Antibody, Detection Antibody Diluent, Sample Diluent, Wash Buffer Concentrate, Conjugate, Conjugate Diluent, Substrate, and Stop Solution.