Order Entry
Czech Republic
ContactUsLinkComponent
PGE2 high sensitivity ELISA kit
PGE2 high sensitivity ELISA kit
Catalog # ENZOADI931001
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  
PGE2 high sensitivity ELISA kit
Catalog # ENZOADI931001
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Assay Type:
    Competitive
  • Host:
  • Primary antibody reactivity:
  • Target protein:
    PGE2
  • Description:
    PGE2 high sensitivity ELISA kit
  • Seq:
    02
  • Environmentally Preferable:
  • Sensitivity:
    8,26 pg/ml (range 7,8 - 1,000 pg/ml)

Specifications

About this item

Ultrasensitive PGE2 ELISA kit for inflammation and eicosanoid research.

For the quantitative determination of PGE2 in culture supernatants, serum, saliva, urine, and whole blood from any species.

The PGE2 high sensitivity EIA kit is a colorimetric competitive enzyme immunoassay kit with results overnight + 1 hour.

Ordering information: Contains GxM IgG Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-762 (1x96 wells).