11-dehydro-TXB2 ELISA Kits
Catalog # ENZOADI900092
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:1 KIT
- Assay Type:Competitive
- Target protein:11-dehydro-TXB2
- Description:11-dehydro-TXB2 ELISA kit
- Seq:02
- Sensitivity:4,31 pg/ml (range 9,8 - 10,000 pg/ml)
Specifications
About this item
For the quantitative determination of 11-dehydro-TXB2 in culture supernatants and urine from any species. Cited samples include serum.
The 11-dehydro-TXB2 EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 3 hours. Absorbance is read at 405 nm. This kit offers ultrasensitive quantification of 11-d-TXB2 in a variety of sample matrices with negligible reactivity with related molecules in a convenient 96-well format offers higher throughput analysis compared to conventional analytical methods.
Ordering information: Contains GxR IgG Microtiter plate, Assay buffer, Conjugate, Antibody, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-324 (5x96 wells)