Order Entry
Czech Republic
ContactUsLinkComponent
Leptin (mouse), ELISA kit
Leptin (mouse), ELISA kit
Catalog # ENZOADI900019A
Supplier:  Enzo Life Sciences
CAS Number:  
Leptin (mouse), ELISA kit
Catalog # ENZOADI900019A
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    Leptin (LEP)
  • Description:
    Leptin (mouse), ELISA kit
  • Seq:
    02
  • Sensitivity:
    25,4 pg/ml (range 50 - 3200 pg/ml)

Specifications

About this item

For the quantitative determination of mouse leptin in culture supernatants, plasma, and serum. Cited sample type includes tissue.

The Leptin (mouse), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 3 hours. Absorbance is read at 450nm. The broad dynamic range is suitable for many normal and pathological samples with sensitive quantification of < 26pg/ml mouse leptin.

Ordering information: Contains Microtiter plate, Standard, Antibody, Conjugate, Assay buffer 28, TMB Substrate, Stop solution 2, Wash buffer concentrate.