Zadání objednávky
Czech Republic
ContactUsLinkComponent
ADMA direct (mouse/rat) ELISA kit
  ENZOALX850327KI01
 :  
ADMA direct (mouse/rat) ELISA kit
  ENZOALX850327KI01
 :  
  • Bal.:
    1 KIT
  • Typ testu:
    Direct
  • Primary antibody reactivity:
    Rat
    Mouse
  • Cíl:
    ADMA
  • Popis:
    ADMA direct (mouse/rat) ELISA kit
  • Sekvence:
    02
  • Podmínky uskladnění:
    4 °C (kit), −20 °C (control and standard)
  • Citlivost:
    0,05 µmol/L
For the quantitative detection of asymmetrical dimethylarginine (ADMA) in mouse or rat EDTA-plasma and serum. Does not cross-react with SDMA, NMMA or L-arginine.