Microcystins (adda specific) ELISA kit
Catalog # ENZOALX850319KI01
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:1 KIT
- Description:Microcystins (Adda Specific) ELISA kit
- Seq:02
- Storage conditions:4 °C (long term)
- Sensitivity:0,1 ng/ml (Range 0,15 - 5 ng/ml)
Specifications
About this item
Polyclonal antibody prepared against the Adda moiety binds to microcystins and nodularins, allowing the congener-independent determination of these toxins and many of its congeners, and does not cross-react with other non-related toxins or compounds. No pre-sample preparation required. Enables simultaneous measurement of multiple samples at reasonable costs.
- Convenient – no pre-sample preparation required
- Rapid – analyze 41 duplicate samples in <2,5 hours
- Assay Time: 1 hour 90 minutes