Anti-V5-tag Mouse Monoclonal Antibody (Biotin) [clone: E10/V4RR]
ENZOENZABS2880050
- Antibody type:Primary
- Antigen symbol:V5-tag
- Clonality:Monoclonal
- Clone:E10/V4RR
- Conjugation:Biotin
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Seq:02
- Form:Liquid
- Storage buffer:In PBS containing a proprietary stabilizer and 0,02% sodium azide
- Storage temperature:4 °C
- Shipping temperature:Blue ice not frozen
- Immunogen:Synthetic peptide corresponding to GKPIPNPLLGLDST (V5), conjugated to KLH
- Purification:Purified
- Pk:50 µl
Recommended Applications: ELISA, ICC, IF, IP, WB
Species reactivity: Species independent
Type: Primary
Antigen: V5-tag
Clonality: Monoclonal
Clone: E10/V4RR
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human