Order Entry
Czech Republic
ContactUsLinkComponent
Anti-V5-tag Mouse Monoclonal Antibody (HRP) [clone: E10/V4RR]
Anti-V5-tag Mouse Monoclonal Antibody (HRP) [clone: E10/V4RR]
Catalog # ENZOENZABS2870050
Supplier:  ENZO LIFE SCIENCES
Anti-V5-tag Mouse Monoclonal Antibody (HRP) [clone: E10/V4RR]
Catalog # ENZOENZABS2870050
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    V5-tag
  • Clonality:
    Monoclonal
  • Clone:
    E10/V4RR
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Seq:
    02
  • Form:
    Liquid
  • Storage buffer:
    In PBS containing a proprietary stabilizer and no preservative
  • Storage temperature:
    4 °C
  • Shipping temperature:
    Blue ice not frozen
  • Immunogen:
    Synthetic peptide corresponding to GKPIPNPLLGLDST (V5), conjugated to KLH
  • Purification:
    Purified
  • Pk:
    50 µl

Specifications

About this item

Recommended Applications: WB

Species reactivity: Species independent

Type: Primary
Antigen: V5-tag
Clonality: Monoclonal
Clone: E10/V4RR
Conjugation: HRP
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human