Order Entry
Czech Republic
ContactUsLinkComponent
Anti-V5-tag Rabbit Polyclonal Antibody
Anti-V5-tag Rabbit Polyclonal Antibody
Catalog # ENZOENZABS2860100
Supplier:  ENZO LIFE SCIENCES
Anti-V5-tag Rabbit Polyclonal Antibody
Catalog # ENZOENZABS2860100
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    V5-tag
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG1
  • Western blot:
    Yes
  • Seq:
    02
  • Form:
    Liquid
  • Storage buffer:
    In PBS containing 1 mg/ml BSA and 0,05% sodium azide
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    Synthetic peptide corresponding to GKPIPNPLLGLDST
  • Purification:
    Antigen affinity chromatography
  • Pk:
    100 µG

Specifications

About this item

Recommended Applications: WB

Species reactivity: Species independent

Type: Primary
Antigen: V5-tag
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse