Anti-CCL2 Mouse Monoclonal Antibody (Biotin) [clone: E10052]
Catalog # ENZOENZABS2810100
Supplier: ENZO LIFE SCIENCES
Specifications
- Antibody type:Primary
- Antigen name:C-C motif chemokine 2
- Antigen symbol:CCL2
- Clonality:Monoclonal
- Clone:E10052
- Conjugation:Biotin
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Seq:02
- Form:Liquid
- Antigen synonyms:CCL2|Monocyte chemoattractant protein 1|Small-inducible cytokine A2
- Storage buffer:In PBS containing no preservative
- Storage temperature:–20 °C
- Shipping temperature:Shipped on blue ice
- Immunogen:Full-length recombinant human MCP-1
- Purification:Protein G affinity purified
- Pk:100 µG
Specifications
About this item
Recommended Applications: ELISA, WB
Species reactivity: Human
Type: Primary
Antigen: CCL2
Clonality: Monoclonal
Clone: E10052
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human