Anti-CXCL11 Mouse Monoclonal Antibody (Biotin) [clone: 22F1]
ENZOENZABS2750100
- Antibody type:Primary
- Antigen name:CXCL11
- Antigen symbol:CXCL11
- Clonality:Monoclonal
- Clone:22F1
- Conjugation:Biotin
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Seq:02
- Form:Liquid
- Antigen synonyms:Interferon-γ-inducible protein 9|H174|Beta-R1|Small-inducible cytokine B11|C-X-C motif chemokine 11|Interferon-inducible T-cell α chemoattractant|I-TAC|IP-9|CXCL11
- Storage buffer:In PBS containing 4% BSA and a proprietary preservative
- Storage temperature:–20 °C
- Shipping temperature:Shipped on blue ice
- Immunogen:Full length recombinant ITAC produced in E. coli
- Purification:Protein G affinity purified
- Pk:100 µG
Recommended Applications: ELISA, WB
Species reactivity: Human
Type: Primary
Antigen: CXCL11
Clonality: Monoclonal
Clone: 22F1
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human