Anti-Host cell proteins Rabbit Polyclonal Antibody
ENZOENZABS2630100
- Antibody type:Primary
- Antigen name:Host cell proteins
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Hamster
- Western blot:Yes
- Seq:02
- Form:Liquid
- Antigen synonyms:HCP
- Storage buffer:In PBST with 1% BSA containing 10 ppm Kathon
- Storage temperature:–20 °C
- Shipping temperature:Shipped on blue ice
- Immunogen:E. coli host cell proteins
- Purification:Protein A affinity purified
- Pk:100 µG
Recommended Applications: WB
Species reactivity: E. coli
Type: Primary
Antigen: Host cell proteins
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Hamster