Anti-LAMP3 Mouse Monoclonal Antibody (Alexa Fluor® 546) [clone: 104G4]
Catalog # ENZOENZABS1850100
Supplier: ENZO LIFE SCIENCES
Specifications
- Antibody type:Primary
- Antigen symbol:LAMP3
- Clonality:Monoclonal
- Clone:104G4
- Conjugation:Alexa Fluor® 546
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Rodent,Human,Rat,Mouse
- Seq:02
- Form:Liquid
- Antigen synonyms:DC LAMP|CD208|Lysosome-associated membrane glycoprotein 3|Protein TSC403
- Storage buffer:In PBS containing 50% glycerol
- Storage temperature:−80 °C
- Shipping temperature:Shipped on blue ice
- Immunogen:Human DC-LAMP alkaline phosphatase fusion protein.
- Purification:Purified using QMA Hyper D ion exchange chromatography
- Size:100 µl
- Pk:100 µG
Specifications
About this item
Recommended Applications: IF, IHC (FS)
Species reactivity: Human, Sheep
Type: Primary
Antigen: LAMP3
Clonality: Monoclonal
Clone: 104G4
Conjugation: Alexa Fluor® 546
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Rodent