Anti-HTT Rabbit Polyclonal Antibody
Recommended Applications: WB
Species reactivity: Mouse, Rat
Type: Primary
Antigen: HTT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat
- Catalog No:
- ENZOBMLPW0595A0025
- ENZOBMLPW0595A0100
- Antigen symbol:
- HTT
- HTT
- Antigen name:
- Huntingtin
- Huntingtin
- Antigen synonyms:
- HD Protein
- HD Protein
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Host:
- Rabbit
- Rabbit
- Western blot:
- Yes
- Yes
- Immunogen:
- Synthetic peptide corresponding to aa 2-17 of human huntingtin
- Synthetic peptide corresponding to aa 2-17 of human huntingtin
- Form:
- Liquid
- Liquid
- Purification:
- Protein A affinity purified
- Protein A affinity purified
- Storage buffer:
- In PBS containing 50% glycerol and 0,09% sodium azide
- In PBS containing 50% glycerol and 0,09% sodium azide
- Storage temperature:
- –20 °C
- –20 °C
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together