Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Proteasome 19S Rpn10/S5a subunit Rabbit Polyclonal Antibody
Anti-Proteasome 19S Rpn10/S5a subunit Rabbit Polyclonal Antibody
Supplier:  ENZO LIFE SCIENCES
Certificates
Anti-Proteasome 19S Rpn10/S5a subunit Rabbit Polyclonal Antibody
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Catalog No:
  • ENZOBMLPW10450100
  • ENZOBMLPW10450025
  • Antigen symbol:
  • Proteasome 19S Rpn10/S5a
  • Proteasome 19S Rpn10/S5a
  • Antigen name:
  • Proteasome 19S Rpn10/S5a Subunit
  • Proteasome 19S Rpn10/S5a Subunit
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Western blot:
  • Yes
  • Yes
  • Immunogen:
  • Full-length recombinant GST-tagged human S5a (Accession number: P55036)
  • Full-length recombinant GST-tagged human S5a (Accession number: P55036)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Purified by immunodepletion of GST-reactive antibodies followed by Protein A purification
  • Purified by immunodepletion of GST-reactive antibodies followed by Protein A purification
  • Storage buffer:
  • PBS containing 0,01% sodium azide
  • PBS containing 0,01% sodium azide
  • Storage temperature:
  • –20 °C
  • –20 °C
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together