Anti-Proteasome 20S beta5t subunit Mouse Monoclonal Antibody [clone: b8-17]
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Type: Primary
Antigen: Proteasome 20S beta5t subunit
Clonality: Monoclonal
Clone: b8-17
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse
Specifications
- Catalog No:
- ENZOBMLPW10200100
- ENZOBMLPW10200025
- Antigen symbol:
- Proteasome 20S B5T
- Proteasome 20S B5T
- Antigen name:
- Proteasome 20S beta5t Subunit
- Proteasome 20S beta5t Subunit
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- b8-17
- b8-17
- Reactivity:
- Human, Mouse
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- Western blot:
- Yes
- Yes
- Immunogen:
- Recombinant human 20S proteasome beta5t subunit
- Recombinant human 20S proteasome beta5t subunit
- Form:
- Liquid
- Liquid
- Purification:
- Purified by ion exchange chromatography
- Purified by ion exchange chromatography
- Storage buffer:
- PBS containing 0,01% sodium azide
- PBS containing 0,01% sodium azide
- Storage temperature:
- –20 °C
- –20 °C
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together