Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-IgG1 Mouse Monoclonal Antibody [clone: MOPC-21]
 
Anti-IgG1 Mouse Monoclonal Antibody [clone: MOPC-21]

 

  • Catalog No:
  • ENZOADISAB600FI050
  • ENZOADISAB600FI200
  • Antigen symbol:
  • IgG1
  • IgG1
  • Antigen name:
  • Immunoglobulin G1
  • Immunoglobulin G1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • Unconjugated
  • Clone:
  • MOPC-21
  • MOPC-21
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1
  • IgG1
  • Flow cytometry:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Purified by Protein G affinity chromatography
  • Purified by Protein G affinity chromatography
  • Storage buffer:
  • PBS, pH 7,2, containing 0,09% sodium azide
  • PBS, pH 7,2, containing 0,09% sodium azide
  • Storage temperature:
  • 4 °C
  • 4 °C
  • Shipping temperature:
  • Blue ice not frozen
  • Blue ice not frozen

 

Frequently Bought Together