Order Entry
Czech Republic
ContactUsLinkComponent
Anti-14-3-3 eta Rabbit Polyclonal Antibody
Anti-14-3-3 eta Rabbit Polyclonal Antibody
Catalog # ENZOBMLSA4760100
Supplier:  Enzo Life Sciences
Anti-14-3-3 eta Rabbit Polyclonal Antibody
Catalog # ENZOBMLSA4760100
Supplier:  Enzo Life Sciences

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    14-3-3 eta
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Seq:
    02
  • Pk:
    100 µl

Specifications

About this item

Host: Rabbit

  • For Immunohistochemistry, Western Blot

Recognizes human, mouse, rat, cow, sheep and chicken 14-3-3eta.

Type: Primary
Antigen: 14-3-3 eta
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human