Anti-Proteasome 19S Rpt1/S7 Subunit Mouse Monoclonal Antibody
Catalog # ENZOBMLPW94000025
Supplier: Enzo Life Sciences
Specifications
- Antibody type:Primary
- Antigen name:Proteasome 19S Rpt1/S7 Subunit
- Antigen symbol:Proteasome 19S Rpt1/S7
- Clonality:Monoclonal
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Seq:02
- Form:Liquid, in TRIS buffer containing 10 mM sodium azide
- Purification:Purified
- Pk:25 µl
Specifications
About this item
Host: Mouse, Isotype: IgG1
- For Immunoprecipitation
Recognizes human proteasome 26S (19S regulator complex) (Rpt1/S7 subunit).
Type: Primary
Antigen: PROTEASOME 19S RPT1/S7 SUBUNIT
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human