Anti-Proteasome 19S Rpt2/S4 Subunit Rabbit Polyclonal Antibody
Katalogové číslo ENZOBMLPW04450025
Supplier: Enzo Life Sciences
Specifications
- Antibody type:Primary
- Antigen name:Proteasome 19S Rpt2/S4 Subunit
- Antigen symbol:Proteasome 19S Rpt2/S4
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:A. thaliana
- Western blot:Yes
- Seq:02
- Form:Liquid
- Pk:25 µl
Specifications
About this item
Host: Rabbit
- For Western Blot
Recognizes Arabidopsis proteasome 26S (19S regulator complex) (Rpt2a/S4 subunit).
Type: Primary
Antigen: PROTEASOME 19S RPT2/S4 SUBUNIT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Plant (Arabidopsis Thaliana)