Order Entry
Czech Republic
ContactUsLinkComponent
Anti-MMP13 Rabbit Polyclonal Antibody
Anti-MMP13 Rabbit Polyclonal Antibody
Catalog # ENZOALX210833R100
Supplier:  ENZO LIFE SCIENCES
Anti-MMP13 Rabbit Polyclonal Antibody
Catalog # ENZOALX210833R100
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Matrix Metalloproteinase 13
  • Antigen symbol:
    MMP13
  • Clonality:
    Polyclonal
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Seq:
    02
  • Form:
    Liquid. In PBS, pH 7,5, containing 0,01% thimerosal.
  • Antigen synonyms:
    MMP-13|CLG3|Collagenase 3|matrix metallopeptidase 13|MANDP1
  • Concentration:
    1 mg/ml
  • Purification:
    95%
  • Pk:
    100 µl

Specifications

About this item

Host: Rabbit

  • For ELISA, Flow Cytometry, Western Blot

Recognizes human MMP-13. Does not cross-react with human MMP-2, MMP-9, MMP-14, MMP-15, MMP-16, MMP-17 or MMP-24.

Type: Primary
Antigen: MMP13
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human