Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-MMP9 Rabbit Polyclonal Antibody
  ENZOALX210756C100
Anti-MMP9 Rabbit Polyclonal Antibody
  ENZOALX210756C100

 

  • Antibody type:
    Primary
  • Antigen name:
    Matrix Metalloproteinase 9
  • Antigen symbol:
    MMP9
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Western blot:
    Yes
  • Seq:
    02
  • Form:
    Liquid. In PBS containing 0,01% thimerosal.
  • Antigen synonyms:
    92kDa type IV collagenase)|MMP-9|GELB|92 kDa gelatinase|MANDP2|matrix metalloproteinase 9 (gelatinase B|92 kDa type IV collagenase|type V collagenase|matrix metalloproteinase-9|92kDa gelatinase|macrophage gelatinase|CLG4B
  • Concentration:
    1 mg/ml
  • Purification:
    Protein A-affinity purified
  • Pk:
    100 µG

 

 

Host: Rabbit

  • For Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human and marmoset MMP-9. Cross-reacts very slightly with MMP-2. Does not cross-react with collagenases.

Type: Primary
Antigen: MMP9
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human