Anti-HDAC5 Rabbit Polyclonal Antibody
Katalogové číslo ENZOALX210340C100
Supplier: ENZO LIFE SCIENCES
Specifications
- Antibody type:Primary
- Antigen name:Histone Deacetylase 5
- Antigen symbol:HDAC5
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Western blot:Yes
- Seq:02
- ChIP:Yes
- Form:Liquid. In PBS containing 0,05% sodium azide.
- Antigen synonyms:HDAC5|Antigen NY-CO-9|KIAA0600|HD5
- Concentration:0,5 mg/ml
- Purification:Protein G-affinity purified IgG
- Pk:100 µG
Specifications
About this item
Host: Rabbit
- For Immunoprecipitation, Western Blot, ChIP
Recognizes human and mouse HDAC5. Detects a band of`~120kDa.
Type: Primary
Antigen: HDAC5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human