Order Entry
Czech Republic
ContactUsLinkComponent
Anti-IP3 Receptor Type 1 Rabbit Polyclonal Antibody
Anti-IP3 Receptor Type 1 Rabbit Polyclonal Antibody
Catalog # ENZOALX210169R100
Supplier:  ENZO LIFE SCIENCES
Anti-IP3 Receptor Type 1 Rabbit Polyclonal Antibody
Catalog # ENZOALX210169R100
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    IP3 Receptor Type 1
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Western blot:
    Yes
  • Seq:
    02
  • Form:
    Liquid. In PBS containing 1 mg/ml BSA and 0,05% sodium azide.
  • Concentration:
    1,3 mg/ml
  • Purification:
    Epitope-affinity purified IgG
  • Pk:
    100 µl

Specifications

About this item

Host: Rabbit

  • For Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human (weak), mouse, rat and dog IP3R-I. Detects a band of ~240kDa by Western blot.

Type: Primary
Antigen: IP3 RECEPTOR TYPE 1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse