Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-FKBP12 Rabbit Polyclonal Antibody
  ENZOALX210142R100
Anti-FKBP12 Rabbit Polyclonal Antibody
  ENZOALX210142R100

 

  • Antibody type:
    Primary
  • Antigen name:
    FK506 Binding Protein 12
  • Antigen symbol:
    FKBP12
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Western blot:
    Yes
  • Seq:
    02
  • Form:
    Liquid. In PBS containing 1 mg/ml BSA and 0,05% sodium azide.
  • Antigen synonyms:
    FKBP-12|FKBP1|EC 5.2.1.8|12 kDa FKBP|Rotamase|FKBP-1A|FKBP1A|PPIase FKBP1A|Peptidyl-prolyl cis-trans isomerase FKBP1A|FKBP12|Immunophilin FKBP12|FK506-binding protein 1A
  • Concentration:
    1 mg/ml
  • Purification:
    Epitope-affinity purified IgG
  • Pk:
    100 µl

 

 

Host: Rabbit

  • For Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human, mouse and rat FKBP12. Detects a band of ~12kDa by Western blot.

Type: Primary
Antigen: FKBP12
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human