Order Entry
Czech Republic
ContactUsLinkComponent
 
Restriction enzymes, DraI, FastDigest™, Fermentas
Catalog # FD0224
CAS Number:  
undefined
Restriction enzymes, DraI, FastDigest™, Fermentas
Catalog # FD0224
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Compatible buffer:
    FastDigest Green Buffer or FastDigest Buffer (colorless)
  • Overhang type:
    Blunt
  • Description:
    DraI, FastDigest™
  • Enzyme name:
    DraI
  • No. of reactions:
    200 (200 µl)

Specifications

About this item

Thermo Scientific™ FastDigest™ enzymes are an advanced line of restriction enzymes for rapid DNA digestion.

  • 100% activity of all FastDigest™ enzymes in one universal reaction buffer
  • Direct loading on gels with FastDigest™ Green buffer
  • 100% buffer compatibility with downstream applications

Designed to complete digestion in 5 to 15 minutes.