Restriction enzymes, HindIII, FastDigest™, Fermentas
Supplier: Thermo Fisher Scientific
Certificates
About this item
Thermo Scientific™ FastDigest™ enzymes are an advanced line of restriction enzymes for rapid DNA digestion.
- 100% activity of all FastDigest™ enzymes in one universal reaction buffer
- Direct loading on gels with FastDigest™ Green buffer
- 100% buffer compatibility with downstream applications
Designed to complete digestion in 5 to 15 minutes.
Specifications
- Compatible buffer:FastDigest Green Buffer or FastDigest Buffer (colorless)
- Overhang type:5' overhang
- Enzyme name:HindIII