Order Entry
Czech Republic
ContactUsLinkComponent
Restriction enzymes, SfiI, Fermentas
Restriction enzymes, SfiI, Fermentas
Catalog # ER1821
CAS Number:  
undefined
Restriction enzymes, SfiI, Fermentas
Catalog # ER1821
CAS Number:  

Specifications

  • Concentration:
    10 U/µl
  • Pk:
    1 KIT
  • Compatible buffer:
    FastDigest Green Buffer or FastDigest Buffer (colorless)
  • Overhang type:
    3' overhang
  • Description:
    SfiI
  • Enzyme name:
    SfiI
  • Units:
    1000 units

Specifications

About this item

SfiI restriction enzyme recognizes GGCCNNNN^NGGCC sites and cuts best at 50 °C in G buffer.

  • Convenient colour-coded five buffer system
  • Includes universal Tango buffer for double-digestions
  • BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities

Storage buffer: 10 mM Tris-HCl (pH 7,4 at 25 °C), 300 mM NaCl, 10 mM MgCl2, 1 mM DTT, 0,15% triton X-100, 0,2 mg/ml BSA and 50% (v/v) glycerol. After 50-fold overdigestion with SfiI, more than 95% of the DNA fragmentscan be ligated and recut. Minimum 5 units of theenzyme are required forcomplete digestion of 1 µg of agarose-embedded adenovirus-2 DNA in 16 hours.

Applications include molecular cloning, restriction site mapping, genotyping, southern blotting, restriction fragment length polymorphism (RFLP) and SNP.