ContactUsLinkComponent
Combined fridge freezers, PremiumPlus BioFresh, CBNes series
  LIEBCBNES5156
 :  Liebherr
Combined fridge freezers, PremiumPlus BioFresh, CBNes series
  LIEBCBNES5156
 :  Liebherr
  • W×D×H ext.:
    750×630×2020 mm
  • Capacity:
    300 + 115 l
  • Description:
    Combined refrigerator/freezer, SmartSteel coated stainless steel, CBNes 4656, with fan in refrigerator, right interchangeable door hinges
  • Seq:
    02
  • Model:
    CBNes 5156 Premium Biofresh NoFrost
  • Cat. No.:
    LIEBCBNES5156
  • Alarm:
    audible, optical, acoustical
  • Defrost:
    automatic
  • Display:
    LCD display on the door, in the door
  • Fans:
    Fan in refrigerator
  • Material:
    Stainless steel
  • Shelves/drawers:
    7 glass with stainless steel frame shelves (refrigerator), 3 glass drawers (freezer)
  • Specifications:
    Stainless steel slimline door handle with integrated opening mechanism
  • Energy class:
    A+
  • Pk:
    1
These combined fridge freezers with biofresh and NoFrost offer more interior capacity for food storage, economy and energy efficiency.
  • Quiet operation, ease-of-use and exceptional design
  • Sturdy, secure handles have an elegant, premium quality appearance
  • LED ceiling light
  • Features child lock, temperature and door alarm
  • High-performance power cooling system
These freezers feature stainless steel door and solid aluminium door handle. The NoFrost system provides rapid, fan-assisted, frost-free freezing for the safe, longterm storage of food. The NoFrost technology creates a much larger storage capacity and keeps the freezer constantly frost-free. The BioFresh safes pull out on smooth-running telescopic rails and can easily be removed once the door is open to an angle of 90°.
Accurate and independent temperature control of the fridge and freezer compartments is facilitated by DuoCooling: two separately adjustable cooling circuits. There is no air exchange between the fridge and freezer compartment, so that odours cannot be transferred and the stored food is prevented from drying out.
The plumbed-in IceMaker automatically keeps you topped up with the finest quality ice cubes so that you are prepared for every occasion. These freezers feature automatic superfrost function that makes freezing simple and saves energy. It quickly reduces the temperature to –32 °C.