Order Entry
Czech Republic
ContactUsLinkComponent
 
Sodium taurocholate hydrate ≥97.0% (by TLC), Sigma-Aldrich®
 :  Merck
 
Sodium taurocholate hydrate ≥97.0% (by TLC), Sigma-Aldrich®
 :  Merck

 

  • Assay (TLC):
    ≥97,0%
  • Optical activity:
    [α]20/D +23±1°, c= 3% in H₂O (dry matter)
  • Impurities (taurine):
    ≤1%
  • Impurities (cholic acid):
    ≤2%
  • Solubility in H₂O:
    50 mg/ml clear

 

Frequently Bought Together